Learn More
Surgical Design Royaltek™ Stainless Steel Surgical Scalpel Blades
Catalog No. 22079683 Shop All Surgical Design Products
Change view
Click to view available options
Size:
10
11
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-079-683 10
22-079-684 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-079-683 Supplier Surgical Design Supplier No. BLADE#10S
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Surgical Design™ Royaltek™ Stainless Steel Surgical Scalpel Blades are made from sharp, long lasting surgical stainless steel. 10 series blades fit 3, 3L, 3 Graduated, 5B, 7, 9 B3, and B3L scalpel handles. 20 series blades fit 4, 4L, 4 Graduated and 6B.
  • Surgical Design™ presents Royaltek™- our premium line of surgical blades
  • Exceptionally strong and unifoirmly sharpened to maximum acuity
  • Surgical grade stainless-steel blades
  • Individually sealed
  • Peelable packing
  • Sterile
Specifications
Blade Material Stainless Steel
Product Type Scalpel Blade
For Use With (Application) Harvesting Radial Arteries during Coronary Artery bypass Operation, Opening the Bronchus during Thoracic Surgery, Inguinal Hernia Repair
Size 10
For Use With (Equipment) Fits Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L.
Description Primarily used for Small Inscisions in Skin and Muscle
Sterility Sterile