Learn More
Dojindo Molecular Technologies Inc ROS Assay Kit -Highly Sensitive DCFH-DA- (100 tests) allows high-sensitivity detection of total ROS with fluorescence microscopes, plate readers or flow cytometers (Ex: 505 nm/Em: 525 nm) 100 tests
Supplier: Dojindo Molecular Technologies Inc R25210
Reactive oxygen species (ROS) are primarily produced in mitochondria during ATP synthesis. ROS play an essential role in signaling pathways and the immune system, whereas increasing ROS is associated with diseases and cellular senescence. Recent studies suggested that ferroptosis is a new type of cell death characterized by iron dependency and increasing ROS. Thus, ROS detection has been attracting considerable interest in ferroptosis research. DCFH-DA is widely used for ROS detection, but it has some limitations like weak fluorescence signals and high background. Dojindo’s ROS Assay Kit -Highly Sensitive DCFH-DA- overcomes these limitations. The dye employed in the kit allows ROS detection with higher sensitivity than DCFH-DA. Moreover, the Loading Buffer included in this kit maintains cellular health during assays. Dojindo has various reagents for ferroptosis research. (https://www.dojindo.com/ferroptosis-detection-selection-guide/)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.