Learn More
Vector Laboratories Peanut Agglutinin (PNA) Rhodamine-labeled 5 mg
SDP
Supplier:  Vector Laboratories RL10725
Encompass
Peanut agglutinin (PNA) binds the T-antigen, a galactosyl (β-1,3) N-acetylgalactosamine structure present in many glycoconjugates and other soluble and membrane-associated glycoproteins and glycolipids. With exceptions, the PNA receptor sequence is normally sialylated, which prevents binding to its receptor oligosaccharide (see Jacalin). Sialic acid not bound to receptor sugars may also inhibit binding, and including calcium in diluents can enhance binding. This rhodamine-labeled PNA features a ratio of fluorophores to lectin protein that provides optimal staining (excitation 575 nm, emission 600 nm). It is supplied as a solution essentially free of unconjugated fluorophores.
Catalog No. NC9604017
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.