Learn More
BD Rhapsody™ Cartridge Kit
Catalog No. BDB633733 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
BDB633733 1 Ea.
1 options
This item is not returnable. View return policy
Catalog No. BDB633733
Supplier: BD 633733
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
This kit contains four individually packed single-use BD Rhapsody™ Cartridges that work with the BD Rhapsody™ System.
  • Contains four individually packed single-use BD Rhapsody™ Cartridges that work with the BD Rhapsody™ System
  • Utilizing the gentle, gravity-based, microwell technology, these cartridges help you to capture single cells from your samples
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.