Learn More
Hach Company Nitrogen-Ammonia Reagent Set, TNT, AmVer (Salicylate), High Range
Supplier:  Hach Company 2606945
Encompass
For determination of High Range Ammonia Nitrogen by the AmVer™ Salicylate Test 'N Tube™ Method. Hach Method 10031. Range: 0.4 - 50.0 mg/L NH3-N. Set contains AmVer™ Diluent Reagent High Range Test 'N Tube™, Ammonia Salicylate Reagent Powder Pillows and Ammonia Cyanurate Reagent Powder Pillows. 25-50 tests (dependent upon the number of reagent blank tests performed).
Catalog No. 50-199-6332