Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RERE Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS7077R
Description
Required for vesicle-mediated transport. Catalyzes the fusion of transport vesicles within the Golgi cisternae. Is also required for transport from the endoplasmic reticulum to the Golgi stack. Seem to function as a fusion protein required for the delivery of cargo proteins to all compartments of the Golgi stack independent of vesicle origin.Specifications
| RERE | |
| Polyclonal | |
| Unconjugated | |
| RERE | |
| 1110033A15Rik; AI414665; ARG; arginine glutamic acid dipeptide (RE) repeats; arginine-glutamic acid dipeptide (RE) repeats; arginine-glutamic acid dipeptide repeats; Arginine-glutamic acid dipeptide repeats protein; Arp; ATN1L; Atr2; atrophin 2; atrophin-1 like protein; atrophin-1 related protein; Atrophin-1-like protein; Atrophin-1-related protein; Atrophin-2; AW742570; DNB1; eye |
|
| Rabbit | |
| Protein A | |
| RUO | |
| 473 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
| Q9P2R6 | |
| RERE | |
| KLH conjugated synthetic peptide derived from human RERE. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction