Learn More
Dentec Safety™ Replacement Banana Oil
SDP
Catalog No. 19222544 Shop All Dentec Safety Specialists Inc. Products
Click to view available options
:
3.5 oz
This item is not returnable. View return policy
Catalog No. 19-222-544
Supplier: Dentec Safety™ 79241
This item is not returnable. View return policy
Banana oil (isoamyl acetate) has a strong, recognizable odor to quickly indicate the lack of proper respirator fit.
Specifications
For Use With (Equipment) Masks
Model 79241
Quantity 1 Ea.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.