Learn More
Thermo Scientific™ Remel™ Candida albicans Test Kit
Description
Thermo Scientific™ Remel Candida albicans Test Kit is a reagent impregnated disc test used for rapid presumptive identification of Candida albicans.
- Rapid test for the detection of enzymes L-proline aminopeptidase and Β-galactosaminidase in yeast cells for the presumptive identification of Candida albicans cultured on laboratory media
A rapid and sensitive method utilizing a combination of p-nitrophenyl-N-acetyl-Β-D-galactosaminide (NGL) and L-proline-Β-naphthylamide (PRO)
Includes:
50 test discs; 1 × 45mL vial 03% sodium hydroxide; 1 vial color developer; 50 plastic tubes with caps; wooden applicator sticks.
Specifications
Specifications
| Content And Storage | Unopened reagents stable until the expiration date when stored at 2°C to 8°C |
| Description | Candida albicans Test Kit |
| CE Marker | Yes |
| Quantity | 50 Tests |
For in vitro diagnostic use