Learn More
Cole-Parmer Cole-Parmer Fluorescent Fwt Red Dye Tablets, 200 Tablets
Supplier: Cole-Parmer 0029516
Dyes diffuse rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more. Fluorescent red dye (rhodamine WT) is noncarcinogenic. Ideal in yellowed water or water with heavy algae growth. Use an ultraviolet lamp (order separately) to trace fluorescent red dyes where normal visual detection is difficult. Minimum detectable concentration of all dyes in clear water is 1 ppm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.