Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Red Blood Cell (RBC) Lysis Buffer
SDP

Catalog No. NBP229442 Shop All R&D Systems Products
Click to view available options
Quantity:
50 mL

Catalog No. NBP229442

Supplier: Novus Biologicals™ NBP229442

Only null left
Add to Cart
Add to Cart

Applications: Functional

The Red Blood Cell (RBC) Lysis Buffer is for optimal lysis of erythrocytes in single-cell suspensions of mouse hematopoietic tissues such as spleen and human peripheral blood. This buffer contains ammonium chloride, which lyses red cells with minimal effect on lymphocytes. Nucleated red blood cells are not effectively lysed with ammonium chloride. RBC lysis is not necessary when working with mouse thymus and lymph node. Buffer: Dilute 10X RBC Lysis Buffer to 1X in deionized water before use (1mL of 10X RBC lysis buffer with 9mL of deionized water). Bring to room temperature prior to use. Reconstitute: Solubilize the peptides prior to use by making 5mM PBS stock solutions

Specifications

Concentration Functional
For Use With (Application) Use in functional reported in scientific literature (PMID: 24371146)
Quantity 50 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.