Learn More
Promega Reagent Powder for CellTiter 96™ AQueous Nonradioactive Cell Proliferation Assay Kit (MTS)
Shop All Promega Corporation ProductsDescription
- A Novel Tetrazolium Compound for Use in Colorimetric Assays
- Available in two sizes
- Use in assays to determine the number of viable cells
Specifications
Specifications
| Quantity | 250 mg |
| Detection Method | Colorimetric |
| For Use With (Application) | Determining the number of viable cells in proliferation |
| Content And Storage | -20°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.