Learn More
Ace Glass, Inc. REACTOR BODY ONLY REACTOR BODY ONLY
Supplier: Ace Glass, Inc. 788020
Borosilicate reactor body with #25 Ace-Thred at top and 14/10 outer side joint externally threaded to allow an o-ring cap seal connection with mating inner joint. Also featured is a porosity E (4-8m) filter disc at the bottom to be used as a gas inlet to help remove oxygen from the solution and/ or for mixing.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.