Learn More
United Scientific™ Rain Gauge
SDPSCI_ED
Catalog No. S00330 Shop All United Scientific Supplies, Inc Products
Click to view available options
:
Rain Gauge
This item is not returnable. View return policy
Catalog No. S00330
Supplier: United Scientific™ UNRG001
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Portable rain gauge can be mounted on a wall or staked into the ground.
  • The gauge measures up to 5 in. of rain fall
  • Made from durable polystyrene
  • The large clear increments are easy to read and the curved lens magnifies the look of the numbers and markings when rain is captured
  • Product dimensions (L x W x D) are 12 x 2.7 x 1.5 in.
Specifications
Product Type Rain Gauge
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.