Learn More
RASA2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RASA2.
Supplier: Abnova Corporation PAB22679
Description
The protein encoded by this gene is member of the GAP1 family of GTPase-activating proteins. The gene product stimulates the GTPase activity of normal RAS p21 but not its oncogenic counterpart. Acting as a suppressor of RAS function, the protein enhances the weak intrinsic GTPase activity of RAS proteins resulting in the inactive GDP-bound form of RAS, thereby allowing control of cellular proliferation and differentiation. This particular family member has a perinuclear localization and is an inositol 1,3,4,5-tetrakisphosphate-binding protein; a compound suggested to function as a second messenger. [provided by RefSeq
Sequence: PDDYSNFVIEDSVTTFKTIQQIKSIIEKLDEPHEKYRKKRSSSAKYGSKENPIVGKASSpecifications
| RASA2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RASA2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| GAP1M | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5922 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| RASA2 | |
| RASA2 | |
| Recombinant protein corresponding to amino acids of human RASA2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only