Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Phospho-Ubiquitin (S65), Polyclonal, R&D Systems™

Catalog No. A110
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A110 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A110 Supplier R&D Systems Supplier No. A110
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Ubiquitin Polyclonal specifically detects Ubiquitin in Human samples. It is validated for Western Blot.

Specifications

Antigen Ubiquitin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.5-1 ug/mL
Formulation 0.5 mg/ml in PBS pH 7.4, 50% (v/v) Glycerol with No Preservative
Gene Accession No. P0CG47
Gene Alias RPS27A, UBA52, UBB, UBB ubiquitin B, UBC
Gene Symbols UBB
Host Species Rabbit
Immunogen Peptide sequence from Ubiquitin, phosphorylated at Serine 65. Accession # P0CG47
Purification Method Antigen Affinity-purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6233
Test Specificity This antibody detects mono- and poly-Ubiquitin chains that are phosphorylated on Serine 65. It has no cross-reactivity with non-phosphorylated Ubiquitin, Ubiquitin phosphorylated at Serine 57 or Tyrosine 59, or phosphorylated Parkin. Detects only recombinant, phosporylated Ubiquitin. Not recommended on natural lysates (see western blot results below).
Target Species Human
Content And Storage 6 months from date of receipt, -20 degreesC as supplied. 3 months, -20 degreesC under sterile conditions after opening.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.