Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[(R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II) Dichloride, TCI America™
SDP

Catalog No. B3067250MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
250 mg

Catalog No. B3067250MG

Supplier: TCI America B3067250MG

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 132071-87-5
Molecular Formula C44H32Cl2P2Ru
Molecular Weight (g/mol) 794.66
MDL Number MFCD01073794
InChI Key YEKBVMDAGDTOQB-UHFFFAOYSA-L
Synonym Dichloro[(R)-2,2′C-bis(diphenylphosphino)-1,1′C-binaphthyl]ruthenium(II)
PubChem CID 11136527
IUPAC Name [2'-(diphenylphosphanyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphane; dichlororuthenium
SMILES Cl[Ru]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 250 mg
Formula Weight 794.66
Physical Form Crystalline Powder
Chemical Name or Material [(R)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthyl]ruthenium(II) Dichloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.