Learn More
TARGETMOL CHEMICALS INC QX-314 CHLORIDE 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T12608100MG
Encompass_Preferred
Also available in 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. QX-314 chloride is a membrane-impermeable permanently charged blocker of sodium channel. purity: 98%
Catalog No. 50-257-7421
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.