Learn More
New England Biolabs, Inc. Quick CIP – 1000 units
Supplier: New England Biolabs, Inc. M0525S
Quick CIP is a heat-labile recombinant version of calf intestinal alkaline phosphatase (CIP) purified from a recombinant source. Quick CIP nonspecifically catalyzes the dephosphorylation of 5 and 3 ends of DNA and RNA phosphomonoesters. It also hydrolyses ribo- and deoxyribonucleoside triphosphates (NTPs and dNTPs). Quick CIP is useful in many applications that require the dephosphorylation of DNA or RNA ends. In cloning, dephosphorylation prevents re-ligation of linearized plasmid DNA. The enzyme can quickly dephosphorylate 5 protruding, 5 recessed, and blunt ends in just 10 minutes. Quick CIP may also be used to degrade unincorporated dNTPs in PCR reactions to prepare templates for DNA sequencing or SNP analysis.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.