Learn More
Agilent Technologies QUICK CHANGE II-E SITE (10RXN)
Supplier: Agilent Technologies 200555
QuikChange II-E (Electroporation) Site; Directed Mutagenesis Kit; Highest Accuracy; Site Directed Mutagenesis Method; Fast; Contains PfuUltra High Fidelity DNA polymerase, dNTP mix; 1/KT (10 rxn)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.