Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Qubit™ 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits
GREENER_CHOICE

Catalog No. Q33231
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 assays
500 assays
Assay:
1X dsDNA, broad range
1X dsDNA, high sensitivity
Quantitation Range:
0.1 to 120 ng
4 to 4000 ng
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Assay Quantitation Range
Q33231 500 assays 1X dsDNA, high sensitivity 0.1 to 120 ng
Q33230 100 assays 1X dsDNA, high sensitivity 0.1 to 120 ng
Q33266 500 assays 1X dsDNA, broad range 4 to 4000 ng
Q33265 100 assays 1X dsDNA, broad range 4 to 4000 ng
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. Q33231 Supplier Invitrogen™ Supplier No. Q33231
Only null left
Add to Cart
Edge
Add to Cart

Highly selective for dsDNA over RNA, the Qubit 1X dsDNA HS and BR Assay Kits offer easy-to-use formulations for the accurate quantitation of dsDNA in 100 and 500 assay formats. Lambda Standards provide sufficient material for up to 500 samples.

Experience easy, accurate, and precise quantification of dsDNA with Qubit 1X dsDNA HS (High Sensitivity) and Qubit 1X dsDNA BR (Broad Range) Assay Kits. The 1X formulation provides a ready-to-use working solution that enables simple dsDNA sample quantification on Qubit Fluorometers. These assays are highly selective for dsDNA over ssDNA, RNA, protein, and free nucleotides. Contaminants, such as salts, solvents, or detergents are well-tolerated.

Qubit 1X dsDNA HS and BR Assay Kits, designed for use with Qubit Fluorometers, are highly selective for double-stranded DNA (dsDNA) over single-stranded DNA (ssDNA), RNA, protein, and free nucleotides. All kits provide a ready-to-use working solution and DNA standards. To perform the assay, simply dilute your sample (any volume from 1–20 μL is acceptable) into the 1X working solution provided, then read the concentration using a Qubit Fluorometer.

Qubit 1X dsDNA HS Assay Kit
The Qubit dsDNA HS (High Sensitivity) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of sensitive DNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial DNA sample concentrations of 5 pg/μL to 120 ng/μL, providing a detection range of 0.1−120 ng.

Qubit 1X dsDNA BR Assay Kit
The Qubit dsDNA BR (Broad-Range) Assay Kit, designed for use with Qubit 4 or Qubit Flex Fluorometers, provides an accurate and selective method for the quantitation of DNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial DNA sample concentrations of 0.2 to 4,000 ng/μL, providing a detection range of 4-4,000 ng.

Notes
• Qubit 1X dsDNA HS can be used with Qubit 2, Qubit 3, Qubit 4, and Qubit Flex Fluorometers
• Qubit 1X dsDNA BR are designed for use with Qubit 4 and Qubit Flex Fluorometers
• Use thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200 μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer

TRUSTED_SUSTAINABILITY

Specifications

Assay 1X dsDNA, high sensitivity
For Use With (Equipment) Qubit 2.0, 3, 4, and Flex Fluorometers
Green Features Fewer resources used, Less waste
No. of Reactions 500 Reactions
Quantitation Range 0.1 to 120 ng
Product Line Qubit
Quantity 500 assays
Shipping Condition Wet Ice
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.