Learn More
Research Products International Corp Pyridoxine Hydrochloride [Vitamin B6], 50 Grams

Supplier: Research Products International Corp P5024050.0
CAS Number: 58-56-0
Molecular Formula: C8H11NO3 • HCl
Molecular Weight: 205.6
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.