Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ UniTranz-RT Transport Systems (Media only)

Catalog No. p-4574234
Click to view available options
Fill Volume:
1 mL
3 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

For the collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma

Intended for the collection and preservation of Viruses - Chlamydia - Mycoplasma and Ureaplasma; Contains antimicrobial agents to minimize bacterial and fungal contamination; Contains glass beads to release and disperse;Used in viral antigen detection test, PCR and rapid tests sample into the medium during vortexing; Available in a 1ml or 3ml fill, medium is stable at room temperature; No swab, media only; Premium medical grade plastic components to improve product shelf life; Shelf life - 18 months; Made in USA

Specifications

Product Type Transport System
Sterility Sterile
Fill Volume 1 mL
For Use With (Application) For the collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
Includes Vial filled with 1mL universal transport medium
Packaging Individually-wrapped
Quantity 300 Cs.
Storage Requirements 2°C to 25°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.