Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Puritan™ UniTranz-RT Transport Systems
Click to view available options
Fill Volume:
1 mL
3 mL
Swab Configuration:
Dual Applicator
Single Applicator
Shaft Material:
Aluminum, Plastic
Polystyrene
Tip Type:
1 Standard, 1 Minitip
Elongated Flocked
Elongated, Ultrafine Flocked
Minitip Flocked
Ultrafine Flocked
Description
For the collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
Intended for the collection and preservation of Viruses - Chlamydia - Mycoplasma and Ureaplasma; Contains antimicrobial agents to minimize bacterial and fungal contamination;Contains glass beads to release and disperse;Used in viral antigen detection test, PCR and rapid tests sample into the medium during vortexing; Used in viral antigen detection test, PCR and rapid tests; Available in a 1ml or 3ml fill, medium is stable at room temperature; Advanced kit design is easy to use and reliable; Self-centering cap is leakproof and facilitates easy swab removal and disposal; High-performance swab tip features patented PurFlock Ultra™ swab with molded breakpoint designed to accommodate different collection sites; Soft tip for patient comfort; Premium medical grade plastic components to improve product shelf life; All kits include sterile wrapped swabs and vial; Shelf life - 18 months; Made in USA
Specifications
Specifications
Product Type | UniTranz-RT Transport System |
Media Type | Hank's Balanced Salts, Bovine Serum Albumin, Gelatin, Sucrose, L-Cysteine, L-Glutamic Acid, Colistin, Amphotericin B, Vancomycin, Sodium Bicarbonate and HEPES Sodium Salt |
Sterility | Sterile |
Swab Configuration | Dual Applicator |
Certifications/Compliance | CE |
Fill Volume | 3 mL |
For Use With (Application) | Collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma |
Includes | One high performance elongated purFlock ultra swab and one ultrafine purFlock ultra swab, wrapped and 3 mL Universal Transport Medium Filled Vial |
Packaging | Individually-wrapped |
pH | 7.3 ±0.2 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction