Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ UniTranz-RT Transport Systems

Catalog No. 22027104
Click to view available options
Fill Volume:
1 mL
3 mL
Swab Configuration:
Dual Applicator
Single Applicator
Shaft Material:
Aluminum, Plastic
Polystyrene
Tip Type:
1 Standard, 1 Minitip
Elongated Flocked
Elongated, Ultrafine Flocked
Minitip Flocked
Ultrafine Flocked
This item is not returnable. View return policy
This item is not returnable. View return policy

For the collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma

Intended for the collection and preservation of Viruses - Chlamydia - Mycoplasma and Ureaplasma; Contains antimicrobial agents to minimize bacterial and fungal contamination;Contains glass beads to release and disperse;Used in viral antigen detection test, PCR and rapid tests sample into the medium during vortexing; Used in viral antigen detection test, PCR and rapid tests; Available in a 1ml or 3ml fill, medium is stable at room temperature; Advanced kit design is easy to use and reliable; Self-centering cap is leakproof and facilitates easy swab removal and disposal; High-performance swab tip features patented PurFlock Ultra™ swab with molded breakpoint designed to accommodate different collection sites; Soft tip for patient comfort; Premium medical grade plastic components to improve product shelf life; All kits include sterile wrapped swabs and vial; Shelf life - 18 months; Made in USA

Specifications

Product Type UniTranz-RT Transport System
Media Type Hank's Balanced Salts, Bovine Serum Albumin, Gelatin, Sucrose, L-Cysteine, L-Glutamic Acid, Colistin, Amphotericin B, Vancomycin, Sodium Bicarbonate and HEPES Sodium Salt
Sterility Sterile
Swab Configuration Single Applicator
Certifications/Compliance CE
Fill Volume 3 mL
For Use With (Application) Collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
Includes High performance mini tip purFlock ultra swab, wrapped and 3 mL Universal Transport Medium Filled Vial
Packaging Individually-wrapped
pH 7.3 ±0.2
Quantity 300 Cs.
Shaft Material Polystyrene
Storage Requirements 2°C to 25°C
Tip Type Minitip Flocked
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.