Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ Advanced Hand Sanitizer Soothing Gel
SDP

Catalog No. 19043906 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Color:
Clear
Green
Packaging:
Portable Flip Cap Bottle
Portable Pump Bottle
Refill
Table Top Pump Bottle
Quantity:
12/Pk.
24/Pk.
8/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Packaging Quantity
19-043-906 Green Refill 8/Pk.
19-043-944 Green Portable Flip Cap Bottle 24/Pk.
19-043-915 Green Table Top Pump Bottle 12/Pk.
19-043-946 Clear Portable Pump Bottle 24/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-043-906 Supplier Gojo Industries Inc Supplier No. 213708
This item is not returnable. View return policy

Contains aloe and vitamin E.

  • Kills most illness causing germs without damaging skin
  • Formulated with moisturizers and skin conditioners to keep hands soft
  • Refill pack option is SANITARY SEALED™ to prevent contamination
TRUSTED_SUSTAINABILITY

Specifications

Product Type Sterilizer/Disinfectant Hand Gel
Color Green
Form Gel
For Use With (Equipment) PURELL™ NXT™ Dispenser
Packaging Refill
Quantity 8/Pk.
Size 1000 mL
Volume (Metric) 1000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.