Learn More
Cary Company Pump Adapter for Rieke Flexspout II
Supplier: Cary Company 30WFSA
This pump adapter is designed to add a hand pump feature to containers with a Rieke Flexspout II. This adapter eliminates the need for tipping to dispense product, which reduces the risk of spills or accidents. This adapter converts 40 mm threads to 38 mm.