Learn More
Enzo Life Sciences Puerarin (5mg). CAS: 3681-99-0
Supplier: Enzo Life Sciences ALX350249M005
Biologically active isoflavone. Affects serotonin levels and platelet aggregation in blood cells. 5-HT2c antagonist. Antibacterial. Displays cardioprotective effects. Induces apoptosis. Alternative name: 8-(beta-D-Glucopyranosyl-7-hydroxy-3- (4-hydroxyphenyl)-4H-1-benzopyran-4-one, NPI-031G. Purity: ≥98% (HPLC). Solubility: Soluble in methanol. Long Term Storage: +4°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.