Learn More
New England Biolabs, Inc. pTXB1 Vector – 10 µg
Supplier: New England Biolabs, Inc. N6707S
pTXB1 is an E. coli expression vector designed for the in-frame insertion of a target gene into the polylinker upstream of the Mxe intein/chitin binding domain. The fusion protein is bound to chitin beads and the thiol-induced cleavage activity of the intein releases the target protein.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.