Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576932
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA576932 100 μL
1 options

Catalog No. PIPA576932

Supplier: Invitrogen™ PA576932

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

IA-2 (insulinoma-associated protein 2, ICA512, PTPRN) is a member of the transmembrane protein tyrosine phosphatase (PTP) family located in secretory granules of neuroendocrine cells. The IA-2 protein is 979 amino acids in length and consists of a luminal domain, transmembrane domain, and cytoplasmic domain. Although a member of the PTP family, IA-2 lacks phosphatase activity with known substrates due to amino acid substitutions at critical sites in its PTP domain. Two paralog RPTPs, IA-2 and IA-2beta were identified as major autoantigens in type-1 diabetes mellitus. On granule exocytosis, the IA-2 cytoplasmic domain is cleaved and the resulting cytosolic fragment moves into the nucleus where it enhances the levels of phosphorylated STAT5 and STAT3, thereby inducing insulin gene transcription and granule biogenesis. IA-2 signaling enhances pancreatic beta-cell proliferation by regulating cyclins D through STATs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRN
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene PTPRN
Gene Accession No. Q16849, Q60673, Q63259
Gene Alias 105 kDa islet cell antigen; BEM-3; brain-enriched membrane-associated protein tyrosine phosphatase; IA2; IA-2; IA-2/PTP; ICA 512; ICA105; ICA3; ICA512; ICA512-CCF; ICA512-cleaved cytosolic fragment; ICA512-N-terminal fragment; ICA512-NTF; ICA512-TMF; ICA512-transmembrane fragment; islet cell antigen 2; islet cell antigen 512; Islet cell autoantigen 3; mIA-A; mKIAA4064; protein tyrosine phosphatase, receptor type N; protein tyrosine phosphatase, receptor type, N; protein tyrosine phosphatase, receptor-type, N; protein tyrosine phosphatase-like N; PTP IA-2; PTPLP; Ptprn; receptor-type tyrosine-protein phosphatase-like N; R-PTP-N
Gene Symbols PTPRN
Host Species Rabbit
Immunogen Recombinant full length Human PTPRN.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116660, 19275, 5798
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.