Learn More
PTF1A Mouse anti-Human, Mouse, Rat, Clone: OTI1H6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810965
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
PFT1A is a component of the pancreas transcription factor 1 complex and is known to have a role in mammalian pancreatic development. The protein plays a role in determining whether cells allocated to the pancreatic buds continue towards pancreatic organogenesis or revert back to duodenal fates. The protein is thought to be involved in the maintece of exocrine pancreas-specific gene expression including elastase 1 and amylase. Mutations in this gene cause cerebellar agenesis and loss of expression is seen in ductal type pancreas cancers.Specifications
| PTF1A | |
| Monoclonal | |
| Unconjugated | |
| PTF1A | |
| PACA, PAGEN2, PTF1-p48, bHLHa29 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 117034, 19213, 256297 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1H6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q64305, Q7RTS3, Q9QX98 | |
| PTF1A | |
| Full length human recombit protein of human PTF1A produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.