Learn More
Agilent Technologies PSHUTTLE-CMV VECTOR
Supplier: Agilent Technologies 240007
pShuttle-CMV Vector contains a multiple cloning site between the CMV promoter and the SV40 polyadenylation signal and is suitable for insertion of a large cDNA (up to 6.6 kb).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.