Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene Glycol, 30% v/v Aqueous, Reagents

Catalog No. 01334510
Click to view available options
:
Quantity: 200 L

Catalog No. 01-334-510

Supplier: Reagents Holdings Llc CP006800200E

Only null left
Add to Cart
Add to Cart

Propylene Glycol, 30% v/v Aqueous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5, 57-55-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym 1,2-Propanediol, Propane-1,2-diol
IUPAC Name water
SMILES O

Specifications

Color Colorless
Boiling Point 102°C
Packaging Poly Drum
Quantity 200 L
Specific Gravity 1.0295
Percent Purity 30%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Propylene Glycol, 30% v/v Aqueous
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.