Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Propylene Glycol 1-Monomethyl Ether 2-Acetate 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
500 mL
Specifications
Specifications
| Melting Point | -67°C |
| Color | Colorless |
| Boiling Point | 146°C |
| Quantity | 500 mL |
| UN Number | 3271 |
| Formula Weight | 132.16 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Propylene Glycol 1-Monomethyl Ether 2-Acetate |
Safety and Handling
| EINECSNumber | (2)-3144 |
| RTECSNumber | AI8925000 |
| TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction