Learn More
Promega P450-Glo CYP3A4 Screening System (Luciferin-PPXE) DMSO-Tolerant Assay
Catalog No. PRV9910 Shop All Promega Corporation Products
Click to view available options
Quantity:
1000 Assays
Catalog No. PRV9910
Supplier: Promega V9910
Only null left
Add to Cart
Add to Cart
Includes
Membrane preparation containing recombinant human cytochrome P450 enzyme; Luminogenic cytochrome P450 substrate appropriate for the enzyme; NADPH Regeneration System; Reaction buffer; Luciferin Detection Reagent; Luciferin-Free Water
The P450-Glo™ CYP3A4 Assay System provides a homogeneous, luminescent method for measuring CYP3A4 activity and the screening system contains a complete set of reagents for performing the assay.
  • Luminescent Assays and Screening Systems for Measuring CYP3A4 Activity
  • Screening systems include complete set of reagents
  • Luminescent format eliminates need for time-consuming analyses
  • Broad dynamic range and low background
Specifications
Quantity 1000 Assays
Content And Storage <-65°C
Product Type P450-Glo™ CYP3A4 Screening System (Luciferin-PPXE) DMSO-Tolerant Assay
The method of recombinant expression of Coleoptera luciferase is covered by U.S. Pat. Nos. 5,583,024; 5,674,713 and 5,700,673
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.