Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega HaloTEV™ Protease For HaloTag™ System

Catalog No. prg6602 Shop All Promega Corporation Products
Click to view available options
Quantity:
4000 U

Catalog No. PRG6602

Supplier: Promega G6602

Only null left
Add to Cart
Add to Cart

HaloTEV Protease (81kDa) is a fusion between the HaloTag™ protein and TEV protease, a highly specific proteolytic enzyme that cleaves at a specific TEV site, a specific seven-amino-acid sequence (ENLYFQ(G/S)).

  • Cleave and Recover Target Protein in a Single Step
  • Easy to covalently remove HaloTEV from your purified protein with HaloLink™ Resin
  • Proteins can be purified tag-free in a single step as the HaloLink™ Resin will bind both HaloTag(R) protein tag and the HaloTEV protease
  • Supplied at a concentration of 5u/μL

Specifications

Concentration 5 U/μL
For Use With (Application) HaloTag™ mammalian Protein purification system
Formulation 1 x 4000U HaloTEV Protease
Content And Storage <-65°C
Quantity 4000 U
Product Type Protease
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.