Learn More
Promega GSK3β Kinase Enzyme System
Catalog No. p-4672007 Shop All Promega Corporation Products
Click to view available options
Quantity:
1/Ea.
10 μg
Catalog No. PRV1991
Supplier: Promega V1991
Only null left
Add to Cart
Add to Cart
Includes
GSK3β Kinase Enzyme System (V1991)
  • Kinase: GSK3β, 10μg (Human, recombinant full length). MW: ∽73kDa
  • Substrate: GSK3 derived from human muscle glycogen synthase 1 (amino acid 636–661)
  • Others: Reaction Buffer, DTT, MnCl
ADP-Glo™ Kinase Assay and EGFR Kinase Enzyme System (V9371)
  • 1 x 10μg GSK3β Kinase Enzyme System (V1991) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)
Recombinant full-length human GSK3 beta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. GSK3 beta is a serine-threonine protein kinase originally identified as the kinase that phosphorylates and inhibits glycogen synthase.
  • Easily Screen and Profile GSK3β Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Content And Storage <-65°C
Quantity 10 μg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.