Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Caspase-Glo™ 8 and Caspase-Glo™ 9 Assay Systems

Catalog No. p-2920758 Shop All Promega Corporation Products
Click to view available options
Quantity:
2.5 mL
10 mL
Product Type:
Caspase-Glo 8 Assay
Caspase-Glo 9 Assay
This item is not returnable. View return policy

Catalog No. PRG8201

Supplier: Promega G8201

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Caspase-Glo 8 Buffer or Caspase-Glo 9 Buffer; Caspase-Glo 8 Substrate or Caspase-Glo 9 Substrate

Caspase-Glo™ Assay is a homogeneous luminescent assay that measures caspase-8 or caspase-9 activity.

  • A Luminescent Assay That Measures Caspase-8 or 9 Activity
  • Decreased assay time and less enzyme needed
  • Homogeneous add-mix-measure protocol amenable to automation

  • U.S. Patent 6,602,677, Australian Patent 754312 and other patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.

Specifications

Quantity 10 mL
Kit Contents 1 x 1 Bottle Caspase-Glo 8 substrate, 1 x 10mL Caspase-Glo 8 Buffer, 1 x 30μL MG-132 inhibitor
Test Time 0.5 to 1 hr.
Product Type Caspase-Glo 8 Assay
Content And Storage -30°C to -10°C
For Use With (Application) Caspase-8 Activation Determination (EC50), Detection of Caspase-8 Activity, Caspase-8 Inhibition Determination (IC50)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.