Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Potassium Polyacrylate
SCI_ED

Catalog No. S25502A Shop All Fisherbrand Products
Click to view available options
:
500g

Catalog No. S25502A

Only null left
Add to Cart
Add to Cart

Specifications

Color White
CAS 25608-12-2
Molecular Formula C6H7KO4
Linear Formula (C3H6O2)n(C3H5KO2)m
Synonym potassium acrylate, potassium polyacrylate, 2-propenoic acid, potassium salt, potassium prop-2-enoate, 79-10-7 parent, polyacrylic acid, potassium salt, 2-propenoic acid, homopolymer, potassium salt, acrylate potassium, acrylic acid homopolymer potassium salt, acmc-20ajv9
Quantity 500 g
InChI Key OESICJXHUIMSJC-UHFFFAOYSA-M
SMILES C=CC(=O)O.C=CC(=O)[O-].[K+]
IUPAC Name potassium;prop-2-enoate;prop-2-enoic acid
Molecular Weight (g/mol) 182.216
PubChem CID 23700835
Packaging Solid
Chemical Name or Material Potassium Polyacrylate
Show More Show Less
ShelfLife 2 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.