Learn More
Research Products International Corp Potassium Chloride, 1 Kilogram
SDP
Supplier:  Research Products International Corp P410001000.0
Encompass_Preferred
Potassium chloride (KCl) is a white, water-soluble salt (74.55 g/mol) used in life science labs to maintain ionic strength and osmotic balance. It supports cell culture, electrophysiology, and buffer preparation, helping regulate ion gradients in biochemical assays, protein purification, and molecular biology workflows requiring controlled potassium levels.
Catalog No. 50-213-627
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.