Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Portable Fuming Chamber

Shop All Sirchie Finger Print Products


Click to view available options
:
Portable Fuming Chamber
Description
Includes
Removable inner shelf with hangers, six evidence clips and black nylon carrying case
- Constructed of transparent, shatter-resistant 0.25 in. acrylic
- Chamber easily assembles in less than five minutes without tools; disassembles into flat sheets and stores in carrying case
- Holds approximately 16 gal., large enough to process multiple items
- Chamber features an inlet compatible with the Portable Humidifier and Cyano-Wand™ (both sold separately)
Specifications
Specifications
| Color | Transparent |
| For Use With (Application) | For cyanoacrylate development in the field |
| Includes | One cna9002 plexiglass fuming chamber with shelf and hangers, one cna9001 black nylon carrying case, six fr1006 evidence clips |
| Material | Shatter-resistant Acrylic |
| Product Type | Portable Fuming Chamber Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction