Learn More
Gibco™ Porcine Serum

Description
- Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays
- It has earned the trust of researchers with consistent quality and award-winning support
- Helps meet research needs and budget requirements
- Sera Category: Specialty
- Origin: New Zealand
- Age: 1 year or less
Order Info
Shipping condition: Frozen (Dry Ice)
Specifications
Specifications
| Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
| Species | Swine/Porcine, Porcine |
| Age | 1 year or less |
| Country of Origin | New Zealand |
| Form | Liquid |
| Product Type | Porcine Serum |
| Quantity | 500 mL |
| Serum Treatment | Standard (Sterile-filtered) |
| Sterility | Sterile |
| Product Line | Gibco |
| Show More |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.