Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Polyvinyl Alcohol Solution, 4%
SCI_ED

Catalog No. S25474 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
Packaging:
Thick Gel - 4% Solution

Catalog No. S25474

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
PubChem CID 11199
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

Physical Form Solid
Linear Formula [CH2CH(OH)]n
Density 1.269g/mL
Quantity 100 mL
Formula Weight 44.05
Packaging Thick Gel - 4% Solution
Chemical Name or Material Polyvinyl Alcohol
ShelfLife 2 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.