Learn More
International Crystal Polystyrene Calibration Film for NIST, PhEur 2.2.24 and PhJap 2.25 Calibration of IR and FTIR Instruments
Supplier: International Crystal 00098448
International Crystal Laboratories' polystyrene calibration film with certificate of traceability to seven (7) NIST frequencies, certified as to absorption minima and maxima differentials at 2870cm-1 and 2849.5cm-1 and 1589cm-1 and 1583cm-1 as required by PhEur 2.2.24 and also certified to PhJap 2.25. Mounted on 2" x 4" card stock.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.