Learn More
Strem, An Ascensus Company CAS# 8017-16-1. 1kg. Polyphosphoric acid (83% P2O5). MFCD00084480
SDP
Supplier:  Strem, An Ascensus Company 9315521000G
Encompass
CAS# 8017-16-1. 1kg. Polyphosphoric acid (83% P2O5). MFCD00084480. Molecular Weight: 239.95. Molecular Formula: H3PO4∙P2O5. Color/form: viscous liq. Strem# 93-1552. http://www.strem.com/catalog/v/93-1552/
Catalog No. 50-901-14227
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.