Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Poly(vinyl chloride), Average Mw 85,000 (Typical), Average Mn 40,000 (Typical), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
25 g
Specifications
Specifications
| CAS | 9002-86-2 |
| Density | 1.4 g/mL (at 25°C) |
| Physical Form | Powder |
| Linear Formula | (CH2 CHCl)n |
| MDL Number | MFCD00084459 |
| Synonym | PVC |
| Type | Analytical Standard |
| Quantity | 25 g |
| Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction