Learn More
Research Products International Corp Replacement Polypropylene Vial Caps, 22mm Metal Foil Lined Plastic Screw Caps, White, 1000 per Package
Supplier: Research Products International Corp 131000
Smooth top white caps available with various type liners for liquid scintillation counting applications. Metal foil liner offers best counting efficiency with excellent sealing properties. Recommended for all samples that have a pH of 5-7. For highly alkaline or acidic samples an inert liner is recommended.
Polypropylene Vial Caps, 22 mm Metal Foil Lined, White, Plastic Screw Cap