Learn More
Bioworld Poly-L-Lysine-Coated Microplates, 50 Plate(s)
Supplier:  Bioworld 201400072
Encompass
Poly-Lysine coated microplatesHigh quality coated microplates.The Poly-Lysine coating of these 96-well microplates offers specific, high affinity binding to a variety of cell types.Clear-colored
Catalog No. 50-197-536
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.