Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMM1/Phosphomannomutase 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$480.74
Specifications
| Antigen | PMM1/Phosphomannomutase 1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
PMM1/Phosphomannomutase 1 Polyclonal specifically detects PMM1/Phosphomannomutase 1 in Human samples. It is validated for Western Blot.Specifications
| PMM1/Phosphomannomutase 1 | |
| Polyclonal | |
| Rabbit | |
| Q92871 | |
| 5372 | |
| Synthetic peptides corresponding to PMM1/Phosphomannomutase 1. The peptide sequence was selected from the N terminal of PMM1/Phosphomannomutase 1. Peptide sequence MAVTAQAARRKERVLCLFDVDGTLTPARQKIDPEVAAFLQKLRSRVQIGV. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| brain glucose-1,6-bisphosphatase, EC 5.4.2.8, phosphomannomutase 1, PMM 1, PMMH22, PMMH-22, Sec53 | |
| PMM1 | |
| IgG | |
| 30 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title