Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Tris/EDTA
Click to view available options
Quantity:
1 L
Description
Thermo Scientific™ Pierce Tris-EDTA (TE) Solution is a sterile reagent for DNA purification and storage.
Additional Information and Online Ordering
Thermo Scientific Pierce Tris-EDTA (TE) Solution is a sterile buffer (pH 8.0) prepared from highly purified reagents and free of DNase activity for DNA purification and storage.
Highlights:
- Prepared with the finest grade chemicals in specially purified water treated to remove particulates, salts, dissolved trace metals, organic compounds and microorganisms
- Autoclave-sterilized and free of DNase contamination
Specifications
Specifications
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C. |
| pH | 8 pH |
| Product Type | Tris/EDTA |
| Quantity | 1 L |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction