Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Streptavidin Magnetic Beads
GSA_VA

Catalog No. PI88816
Click to view available options
Quantity:
1 mL
5 mL
This item is not returnable. View return policy

Catalog No. PI88816

Supplier: Thermo Scientific™ 88816

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Purify or immunoprecipitate biotinylated proteins or affinity ligands using streptavidin-coated magnetic particles for magnet-based separation.

Thermo Scientific Pierce Streptavidin Magnetic Beads accelerate throughput for automated magnetic purification of biotinylated molecules.

Features of Streptavidin Magnetic Beads:

  • High-performance beads—non-aggregating, pre-blocked, iron oxide, superparamagnetic microparticles provide exceptional uniformity for automated HTS and manual applications alike
  • Stable immobilization chemistry—streptavidin is immobilized using leach-resistant chemistry
  • High capacity—superior quality beads with high binding capacity provide rapid and efficient biomolecule purification from complex samples
  • Low non-specific binding—stable, pre-blocked beads provide clean purification products (e.g., antigen eluted in IP with biotinylated antibody) that are compatible with mass spectrometry analysis and other downstream analyses
  • Superior performance—nearly three times higher binding capacity than typical beads from other suppliers, allowing the use of smaller amounts per experiment

Applications:

  • Immunoprecipitate antigens (using biotinylated antibodies) from a wide variety of sources
  • Co-immunoprecipitate interaction complexes using biotinylated antibodies
  • Capture protein-protein interactions in pull-down assays using biotinylated 'bait' proteins
  • Isolate biotin-labeled DNA-protein complexes from cell or tissue extracts
  • Capture single-stranded biotinylated DNA oligos
  • Isolate biotinylated PCR products

These streptavidin magnetic beads are validated and optimized for use with high-throughput magnetic platforms, such as the Thermo Scientific KingFisher 96 and KingFisher Flex Instruments, but the beads also enable premium performance for simple benchtop applications using an appropriate magnetic stand. The iron oxide, super-paramagnetic particles offer superior performance (high capacity and low nonspecific binding) compared with other commercial magnetic beads.

Pierce Streptavidin Magnetic Beads use a recombinant form of streptavidin with a mass of 53kDa and a near-neutral isoelectric point (pI). The protein is a tetramer having four biotin-binding sites. Unlike avidin, streptavidin has no carbohydrate groups, resulting in low nonspecific binding. The high-affinity interaction between streptavidin and biotin cannot be dissociated efficiently except with very harsh conditions, such as boiling in sample loading buffer for SDS-PAGE or 8M guanidine·HCl, pH 1.5. Consequently, it is often possible to elute binding partners in an interaction complex without also eluting the biotinylated component.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1 mL
Concentration Slurry: 10 mg/mL, 1% solids
Content And Storage Upon receipt store at 4°C. Product is shipped on an ice pack.
Particle Size 1 μm
Form Liquid
Product Line Pierce
Quantity 1 mL
Protein Form Recombinant
Target Biotin
Type Magnetic Affinity Bead
Ligand Type Streptavidin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.